Non-sterile PTFE Syringe Filter
Actual product may vary
Fisherbrand
Non-sterile PTFE Syringe Filter
Used for filtration of protein solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore Size
FKS151814890.2 μm
FKS151014990.45 μm
Catalog number FKS15181489
Price (SGD)
82.00
Pack
Pore Size:
0.2 μm
Price (SGD)
82.00
Pack
  • Perfect for daily routine analysis
  • Membrane is made up of PTFE material
  • Low protein binding
  • Hydrophilic membrane
  • Offers medium chemical resistance
Specifications
MaterialPTFE
Material (Membrane)PTFE (Hydrophilic)
Pore Size0.2 μm
SterilityNon-sterile
For Use With (Application)For a broad range of applications
Housing MaterialPolypropylene
Quantity50/Pk.
TypeSyringe Filter
Diameter (Metric)25 mm
Unit SizePack