Protein LoBind™ Tubes
Protein LoBind™ Tubes
Actual product may vary
Protein LoBind™ Tubes
Protein LoBind™ Tubes
Protein LoBind™ Tubes
Fisherbrand
Protein LoBind™ Tubes
Engineered to minimize sample binding and elevate recovery, Eppendorf™ Protein LoBind™ Tubes safeguard your valuable proteins during processing. With cutting-edge LoBind technology, ensure optimal yields for your Western blotting, mass spectrometry, and protein purification applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)PackagingClosure TypeMax. RCF
FKS151783445 mL100 Tubes (2 Bags x 50 Tubes)Snap Cap25,000 x g
AIT00301083025 mL100 Tubes (2 Bags x 50 Tubes)Snap Cap25,000 x g
FIS14-282-3045 mL100 Tubes (2 Bags x 50 Tubes)Snap Cap25,000 x g
Catalog number FKS15178344
Price (SGD)
287.00
PK
Add to cart
Capacity (Metric):
5 mL
Packaging:
100 Tubes (2 Bags x 50 Tubes)
Closure Type:
Snap Cap
Max. RCF:
25,000 x g
Price (SGD)
287.00
PK
Add to cart
  • Surface-coating-free, including substances like silicone, plasticizers, biocides or mold release agents to reduce the risk of sample interference.
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors.
  • Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments.
  • Snap, screw, and SnapTec™ cap options available.
Specifications
Certifications/ComplianceRNase, DNase-free
Closure MaterialHDPE
ColorColorless
Capacity (Metric)5 mL
MaterialPolypropylene
No. per Case100 Cs.
No. per Pack50 Pk.
Packaging100 Tubes (2 Bags x 50 Tubes)
SterilityNon-sterile
AutoclavableAutoclavable
ShapeCylindrical
Capacity (English)0.169 oz.
Bottom ShapeConical
Closure TypeSnap Cap
Writing AreaYes
Product TypeMicrocentrifuge Tube (PCR Clean, Protein LoBind)
Max. RCF25,000 x g
Purity or Quality GradePCR Clean (Non-sterile)
Unit SizePK