Vacuum System
Vacuum System
Actual product may vary
Fisherbrand
Vacuum System
Chemical duty oil-free diaphragm pump designed for chemical duty laboratory applications. Fisherbrand™ Vacuum System is for for evaporation and distillation in combination with rotary evaporators and gel dryers.
Have Questions?
Catalog NumberPump Type
FKS15168886Vacuum System
Catalog number FKS15168886
Price (SGD)
13,886.00
EA
Add to cart
Pump Type:
Vacuum System
Price (SGD)
13,886.00
EA
Add to cart
  • Perfect for evaporation and distillation in combination with rotary evaporators and gel dryers
  • Dry running diaphragm pump and all components coming into contact with vapours are chemically resistant even against acidic, basic and organic solvents
  • Pump heads are made from PTFE with carbon fibre reinforcing to handle corrosive and aggressive vapors without damage
  • Pumping chambers are hermetically separated from drive space, ensuring extended life of mechanical parts
  • Pumps use PFTE diaphragms sandwiched to strong non-wetted backing and PEEK valves, which are chemical resistant and are hard wearing when compared to elastomer valves
  • Gas ballast valve helps to preserve pump performance when working with condensable vapors
  • Addition of digital vacuum gauge and manual regulator allows user to adjust to vacuum level required for their application
  • Inlet separator protects pump for ingress of liquids and particles and exhaust emission condenser protects lab air quality
Specifications
Max. Vacuum8 mbar
Electrical Requirements230 V 50/60 Hz motor with Schuko and UK power leads
Dimensions (L x W x H)36.0 x 31.0 x 44.5 cm (14.1 x 12 x 17.5 in.)
Weight (English)25.4 lb.
Weight (Metric)11.6 kg
Pump TypeVacuum System
Air Displacement16 L/min.
ApplicationsRotary evaporator, Gel dryer, Vacuum oven
Maximum Pressure<8 mbar
Unit SizeEA