Pipette Tips, 50 to 1000 μL
Pipette Tips, 50 to 1000 μL
Actual product may vary
Fisherbrand
Pipette Tips, 50 to 1000 μL
Designed for high-quality micro-pipettes and suitable for various applications. BRAND™ 50 to 1000 μL pipette tips ensure precise and efficient pipetting. Tips are cleanroom-produced, free from contaminants, and environmentally friendly.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackaging
FKS15164897Bulk, 10 x 500
FKS15104897Bulk, 2 x 500
Catalog number FKS15164897
Price (SGD)
33.00
PK
Add to cart
Packaging:
Bulk, 10 x 500
Price (SGD)
33.00
PK
Add to cart
  • Engineered to fit the Transferpette™ line of micropipettes
  • Compatibility tested with other micropipettors from various manufacturers
  • Manufactured and packaged under certified cleanroom conditions according to ISO 14644-1, Class 7 and 8
  • Made from high-quality, pure raw materials, free of DiHEMDA and oleamide
  • No use of mold release agents such as stearates and erucamide in the production process
  • Palletized BIO-CERT™ quality tips are guaranteed to be free from DNA, DNase, RNase, ATP, and endotoxins
  • Thin-walled for high translucency, easy ejection, and low waste
  • Graduations at 250, 500, and 1000 μL for quick volume checks
  • Available either in bulk packaging or convenient, ecological rack-and-stack formats
  • Bulk tips are blue and placed in clear, resealable bags
  • Racked tips are colorless and placed into blue-colored tip trays
  • TipBoxes are made of 100% PP and are fully recyclable
  • The TipRack refilling system reduces waste by more than 20% compared to the TipBox
Specifications
Volume (Metric)50 to 1000 μL
FilteredNon-filtered
SterilityNon-sterile
ColorBlue
AutoclavableAutoclavable
Length (Metric)7 cm
Clean ClaimsDNase, RNase, ATP, and Endotoxin Free
For Use With (Application)For routine laboratory and research pipetting
CompatibilityTransferpette, and most models from Gilson, FINNPIPETTE, Eppendorf, and Sartorius
FormatBulk
Tip StyleUniversal
PackagingBulk, 10 x 500
Product TypePipette Tips
DisposableLimited-use
For Use With (Equipment)Universal Fit
GraduatedYes
MaterialPolypropylene
Graduations250, 500, 1000 μL
Unit SizePK