Non-sterile PTFE Syringe Filter
Actual product may vary
Fisherbrand
Non-sterile PTFE Syringe Filter
Designed for daily routine analysis. Fisherbrand™ Non-sterile PTFE Syringe Filter features a hydrophobic PTFE membrane for filtering HPLC organic solutions. Available in two membrane diameters (13 and 25mm) and two (0.2 and 0.45μm) pore sizes.
Have Questions?
Catalog NumberPore SizeDiameter (Metric)
FKS151514990.45 μm25 mm
Catalog number FKS15151499
Price (SGD)
62.00
Each
Pore Size:
0.45 μm
Diameter (Metric):
25 mm
Price (SGD)
62.00
Each
  • Ideal for filtration of solvents
  • High sample throughput and low protein binding
  • Chemically-resistant hydrophobic PTFE membrane
  • Sturdy polypropylene housing
Specifications
ColorWhite
Material (Membrane)PTFE
Pore Size0.45 μm
SterilityNon-sterile
For Use With (Application)For a broad range of applications
Housing MaterialPolypropylene
Quantity50/Pk.
TypeSyringe Filter
Diameter (Metric)25 mm
Unit SizeEach