Tubes™ with Snap Cap 5.0 mL - Microtube
Tubes™ with Snap Cap 5.0 mL - Microtube
Actual product may vary
Tubes™ with Snap Cap 5.0 mL - Microtube
Tubes™ with Snap Cap 5.0 mL - Microtube
Tubes™ with Snap Cap 5.0 mL - Microtube
Eppendorf Tubes™ with snap caps are your ideal choice when working with medium-sized sample volumes (0.5 mL to 5.0 mL). Designed for user-friendly handling, broad temperature ranges, centrifugation safety (up to 25,000 x g), and sample recovery.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackagingPurity or Quality GradeColor
FKS1514834410 Bags x 20 TubesSterileClear
FIS14-282-3002 Bags x 100 TubesEppendorf QualityClear
FIS14-282-303Individually WrappedBiopur™Clear
Catalog number FKS15148344
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Packaging:
10 Bags x 20 Tubes
Purity or Quality Grade:
Sterile
Color:
Clear
Use these tubes for the simple and safe processing of sample volumes up to 5.0mL. Eppendorf Tubes™ represent the perfect option for working with medium–sized sample volumes.
  • Ideal for working with medium-sized sample volumes ranging from 0.5 mL to 5.0 mL.
  • Designed for ease of use and user-friendly handling during sample preparation and processing.
  • Suitable for a wide range of temperatures, ensuring adaptability to various experimental conditions.
  • Provides reliable safety during centrifugation with a maximum rating of up to 25,000 x g.
  • Specifically designed to enhance sample recovery, preserving the integrity of your samples.
  • Available in Eppendorf Quality, PCR Clean, Biopur™, Sterile, and Forensic DNA Grade quality for tested purity.
  • Free of plasticizers, biocides or mold release agents.
Specifications
Closure MaterialPolypropylene
ColorClear
Capacity (Metric)5 mL
MaterialPolypropylene
No. per Pack200 Pk.
Packaging10 Bags x 20 Tubes
Quantity200 Tubes
SterilitySterile
RNase FreeNo
Dnase FreeNo
AutoclavableAutoclavable
ShapeCylindrical
Capacity (English)0.169 oz.
Clean ClaimsFree of plasticizers, biocides or mold release agents
ACT LabelYes
Bottom ShapeConical
Closure TypeSnap Cap
Closure IncludedYes
Writing AreaYes
Diameter (Metric)17 mm
Product TypeMicrocentrifuge Tube
Max. RCF25,000 x g
Purity or Quality GradeSterile
Unit SizePK