Label Maker Vinyl Cartridges (B-427) for BMP51, BMP53, BMP41 Printers
Label Maker Vinyl Cartridges (B-427) for BMP51, BMP53, BMP41 Printers
Actual product may vary
Fisherbrand
Label Maker Vinyl Cartridges (B-427) for BMP51, BMP53, BMP41 Printers
Designed to adhere to highly curved surfaces, wires and cables
Have Questions?
Catalog NumberColorSize (Metric)Size (English)
FKS15143329Black, White, Clear12.7 x 25.4 mm0.5 x 1 in.
Catalog number FKS15143329
Price (SGD)
252.00
BOX
Add to cart
Color:
Black, White, Clear
Size (Metric):
12.7 x 25.4 mm
Size (English):
0.5 x 1 in.
Price (SGD)
252.00
BOX
Add to cart
  • Feature printable zone in white or yellow
  • Transparent, non printable area wraps around wires and cables for laminating printed texts
  • Offers resistance to abrasion, handling, solvents, water, oil and dirt
  • Available in various sizes
Specifications
ColorBlack, White, Clear
MaterialSelf-laminating Vinyl (B-427)
Product TypeLabel Maker Cartridge
For Use With (Equipment)BMP™51,BMP™53,BMP™41 Printers
Quantity1 Ea. (360 labels)
Size (English)0.5 x 1 in.
Size (Metric)12.7 x 25.4 mm
Width (English)0.5 in.
Width (Metric)12.70 mm
Height (English)1 in.
Height (Metric)25.4 mm
Unit SizeBOX