BMP51/BMP53/BMP41 Label Maker Cartridge: B-422 Polyester
BMP51/BMP53/BMP41 Label Maker Cartridge: B-422 Polyester
Actual product may vary
Fisherbrand
BMP51/BMP53/BMP41 Label Maker Cartridge: B-422 Polyester
For use in commercial and industrial applications
Have Questions?
Catalog NumberDimensions (LxW)
FKS151133291.5 x 0.75 in.
Catalog number FKS15113329
Price (SGD)
222.00
BOX
Add to cart
Dimensions (LxW):
1.5 x 0.75 in.
Price (SGD)
222.00
BOX
Add to cart
  • Permanent adhesive, for powder-coated/textured surfaces
  • Available in various sizes
Specifications
Certifications/CompliancePass 2002/95/EC
DescriptionBMP41/BMP51/BMP53 Labelmaker Labels
Adhesive TypePermanent Acrylic
ColorBlack, White
FinishGloss
MaterialPolyester (B-422)
Product TypeLabel Maker Cartridge
Labels per Row1
Quantity180/Pk.
ShapeRectangle
Size (English)1.5 x 0.75 in.
Size (Metric)38.1 x 19.0 mm
Temperature Range (Metric)-40°C to +100°C
Thickness (Metric)0.102 mm
Dimensions (LxW)1.5 x 0.75 in.
Width (English)1.5 in.
Width (Metric)38.1 mm
Height (English)0.75 in.
Height (Metric)19.0 mm
Unit SizeBOX