Chemgene HLD4L Laboratory Surface Disinfectant
Chemgene HLD4L Laboratory Surface Disinfectant
Actual product may vary
Fisherbrand
Chemgene HLD4L Laboratory Surface Disinfectant
Combines enhanced active ingredient molecules with micelle cleaning technology to provide swift penetration of the cell walls to ensure rapid cell death rather than merely suspending activity.
Have Questions?
Catalog NumberVolume (Metric)
FKS15106686750 mL
Catalog number FKS15106686
Price (SGD)
30.00
EA
Add to cart
Volume (Metric):
750 mL
Price (SGD)
30.00
EA
Add to cart
For multipurpose surface disinfection and cleaning. Chemgene™ Lab Disinfectant Spray destroys bacteria, fungi, mycobacteria, viruses and spores.
Chemgene HLD4L disinfectant concentrate.
  • Cleans and disinfects all areas of the laboratory
  • Manufactured in accordance with Quality Management System ISO 9001
  • Biodegradable under OECD conditions
  • Denatures DNA/RNA
  • Can be used at a dilution of: 1 in 100, General conditions; or 1 in 10 for Bio-hazard contamination
Specifications
Product TypeLab Disinfectant Spray
AppearanceClear
Dilution1 in 100, General conditions; or 1 in 10 for Bio-hazard contamination
For Use With (Application)For multipurpose Surface disinfection and cleaning
Volume (Metric)750 mL
Unit SizeEA