Check Order Status
Non-sterile PTFE Syringe Filter
Actual product may vary
Fisherbrand
Non-sterile PTFE Syringe Filter
Used for filtration of protein solutions.
Have Questions?
Change view
Catalog Number
Pore Size
FKS15101499
0.45 μm
FKS15181489
0.2 μm
2 Options
Catalog number
FKS15101499
Price (SGD)
82.00
PK
Add to cart
Pore Size:
0.45 μm
Price (SGD)
82.00
PK
Add to cart
Perfect for daily routine analysis
Membrane is made up of PTFE material
Low protein binding
Hydrophilic membrane
Offers medium chemical resistance
Specifications
Material
PTFE
Material (Membrane)
PTFE (Hydrophilic)
Pore Size
0.45 μm
Sterility
Non-sterile
For Use With (Application)
For a broad range of applications
Housing Material
Polypropylene
Quantity
50/Pk.
Type
Syringe Filter
Diameter (Metric)
25 mm