AK Latex Cleanroom Gloves
AK Latex Cleanroom Gloves
Actual product may vary
Fisherbrand
AK Latex Cleanroom Gloves
Natural rubber latex chemical resistant gloves feature a diamond embossed palm for secure dry and wet grip. Suitable for Class 100/ISO 5 or higher cleanroom.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FKS1489151110
FKS148715119
Catalog number FKS14891511
Price (SGD)
2,447.00
PK
Add to cart
Size:
10
Price (SGD)
2,447.00
PK
Add to cart
  • 100% natural rubber latex has enhanced dexterity and elasticity
  • Powder free manufacturing process reduces particulate and extractable contamination
  • Diamond embossed palm provides added grip on wet surfaces
  • Cleanroom packaged to protect gloves from airborne contamination
  • Double-bagged with cleanroom compatible packaging and labeling materials
  • Size embossed on gloves so there is no ink to contaminate process
  • Suitable for up to class 100 cleanroom
Specifications
Certifications/ComplianceIEST-RP-CC005.3, ASTM F739
Certifications/Compliance (Europe)EN388, EN374-3
Cleanroom ClassClass 100 (ISO 5)
ColorOrange
Exterior FinishPartially Textured
Latex FreeNo
MaterialLatex
PowderedPowder Free
Size10
SterilityNon-sterile
Thickness (English)20 mil
Thickness (Metric)0.50 mm
ModelAK1815/O/10
SeriesAK
Length (English)15 in.
Length (Metric)38 cm
Unit SizePK