Type 1 Clear Glass Vials, with Dropper Assembly
Type 1 Clear Glass Vials, with Dropper Assembly
Actual product may vary
Type 1 Clear Glass Vials, with Dropper Assembly
Type 1 Clear Glass Vials, with Dropper Assembly
Fisherbrand
Type 1 Clear Glass Vials, with Dropper Assembly
Provides for accurate, reliable dispensing of various samples. Fisherbrand™ Type 1 Clear Glass Vials, with Dropper Assembly are manufactured from clear Type 1 neutral glass.
Have Questions?
Catalog NumberVolume (Metric)
FKS148635725 mL
Catalog number FKS14863572
Price (SGD)
-
Volume (Metric):
5 mL
  • With screw neck
  • With black caps and bulbs
  • Droppers are assembled but not fitted to vials
  • Dropper assembly comprised of clear glass bottle dropper tube fitted to a polypropylene cap with a natural rubber bulb
Specifications
ColorClear
Diameter (Metric) Outer20 mm
MaterialBorosilicate Glass
Volume (Metric)5 mL
Neck StyleScrew
Thread StyleScrew
Closure MaterialPolypropylene
Closure TypeDropper Screw Cap
Height (Metric)43 mm
Quantity40/Pk.
TypeScrew Thread Vial
Unit SizePK