Suitable for the storage of pathology media. Fisherbrand™ Clear Moulded Glass Vials, with Fitted Screw Caps are manufactured from clear Type 1 neutral glass.
Have Questions?
Catalog Number
Volume (Metric)
Closure Material
FKS14863562
30 mL
Polypropylene
Catalog number FKS14863562
Price (SGD)
600.00
PK
Add to cart
Volume (Metric):
30 mL
Closure Material:
Polypropylene
Price (SGD)
600.00
PK
Add to cart
With screw neck
Autoclavable
Available in a range of capacities
With either fitted aluminum rubber lined cap or PP cap