Check Order Status
Soda Lime Glass Vials
Actual product may vary
Fisherbrand
Soda Lime Glass Vials
Use these vials for general mixing and simple laboratory work. Soda Lime Glass Vials will withstand moderate warming (ex. in a water bath).
Have Questions?
Catalog Number
Volume (Metric)
FKS14831582
3 mL
Catalog number
FKS14831582
Price (SGD)
131.00
PK
Add to cart
Volume (Metric):
3 mL
Price (SGD)
131.00
PK
Add to cart
Clear flat bottom vials
Supplied with polythene stoppers
Available in a variety of sizes
Specifications
Diameter (Metric) Inner
12 mm
Material
Soda Lime Glass
Volume (Metric)
3 mL
Includes
Push-in cap
No. per Pack
100
Closure Material
Polyethylene
Closure Type
Push-in Cap
Height (Metric)
38 mm
Quantity
100 Pk.