Bijou Vials with Polypropylene Cap
Bijou Vials with Polypropylene Cap
Actual product may vary
Fisherbrand
Bijou Vials with Polypropylene Cap
Manufactured from Type III Soda Lime Glass with polypropylene cap. Fisherbrand™ Bijou Vials with Polypropylene Cap are suitable for media, diagnostic and sample collection applications.
Have Questions?
Catalog NumberQuantity
FKS14803562245/Pk.
Catalog number FKS14803562
Price (SGD)
218.00
PK
Add to cart
Quantity:
245/Pk.
Price (SGD)
218.00
PK
Add to cart
  • Type III Soda Lime Glass
  • Polypropylene screw cap
  • Clear color bottle
  • Screw thread neck
  • Autoclavable
Specifications
ColorClear
Diameter (Metric) Outer20 mm
MaterialSoda Lime Glass (Type 3)
Volume (Metric)7 mL
For Use With (Application)Suitable for media, diagnostic and sample collection applications
Neck StyleScrew
StyleTubular
Thread StyleScrew
Closure MaterialPolypropylene
AutoclavableAutoclavable
Closure TypeScrew Cap
Height (Metric)43 mm
Quantity245/Pk.
TypeBijou Vial
Closure Size18 mm
Unit SizePK