Check Order Status
Vacuum Filtration And Desiccation Connection Kit
Actual product may vary
Fisherbrand
Vacuum Filtration And Desiccation Connection Kit
Facilitates proper connection to vacuum filtration or dissicator. Welch Ilmvac™ Vacuum Filtration And Desiccation Connection Kit includes a filter and suitable tubing.
Have Questions?
Catalog Number
Includes
FKS13504920
In-Line Hydrophobic Filter, 1.5m Silicone Vacuum Hose (6mm ID) And 1.5m Silicone Vacuum Hose (8mm ID)
Catalog number
FKS13504920
Price (SGD)
434.00
EA
Add to cart
Includes:
In-Line Hydrophobic Filter, 1.5m Silicone Vacuum Hose (6mm ID) And 1.5m Silicone Vacuum Hose (8mm ID)
Price (SGD)
434.00
EA
Add to cart
Quick connection to vacuum filtration funnel / manifold or dessicator
Kit includes in-line hydrophobic filter, 1.5m silicone vacuum hose (6mm I.D.) and 1.5m silicone vacuum hose (8mm I.D.)
Specifications
Includes
In-Line Hydrophobic Filter, 1.5m Silicone Vacuum Hose (6mm ID) And 1.5m Silicone Vacuum Hose (8mm ID)
Quantity
1 Pack
Type
Vacuum Filtration And Desiccation Connection Kit