SP Scienceware™ Break-Safe™ Ampule Opener
SP Scienceware™ Break-Safe™ Ampule Opener
Actual product may vary
SP Scienceware™ Break-Safe™ Ampule Opener
SP Scienceware™ Break-Safe™ Ampule Opener
SP Scienceware™ Break-Safe™ Ampule Opener
SP Scienceware™ Break-Safe™ Ampule Opener
SP Scienceware™ Break-Safe™ Ampule Opener
SP Scienceware™ Break-Safe™ Ampule Opener
SP Scienceware™ Break-Safe™ Ampule Opener
SP Scienceware™ Break-Safe™ Ampule Opener
SP Scienceware™ Break-Safe™ Ampule Opener
SP Scienceware™ Break-Safe™ Ampule Opener
SP Scienceware™ Break-Safe™ Ampule Opener
SP Scienceware™ Break-Safe™ Ampule Opener
Fisherbrand
SP Scienceware™ Break-Safe™ Ampule Opener
Open ampules easily and safely. Bel Art™ SP Scienceware™ Break-Safe™ Ampule Opener allows one handed opening of up to 3 ampules at a time.
Have Questions?
Catalog NumberDimensions (L x W x H)IncludesColor
FKS134924693.75 x 3 x 3.25 in. (9.52 x 7.62 x 8.25cm)4 insert to allow opening of 1, 2, 5 and 10mL ampulesClear
Catalog number FKS13492469
Price (SGD)
313.00
EA
Add to cart
Dimensions (L x W x H):
3.75 x 3 x 3.25 in. (9.52 x 7.62 x 8.25cm)
Includes:
4 insert to allow opening of 1, 2, 5 and 10mL ampules
Color:
Clear
Price (SGD)
313.00
EA
Add to cart
Open ampules easily and safely. Bel Art™ SP Scienceware™ Break-Safe™ Ampule Opener allows one handed opening of up to 3 ampules at a time.
  • Pressing down on the lid snaps ampule necks cleanly
  • Sharp edges are safely contained within the device, away from fingers
  • Includes 4 inserts to allow opening of 1mL, 2mL, 5mL and 10mL ampules
  • Transparency allows unobstructed viewing of ampule contents at all times
  • Copolyester plastic construction has broad chemical resistance
Specifications
ColorClear
Product TypeAmpule Opener
Height (Metric)8.25cm
Height (English)3.25 in.
Length (Metric)9.52cm
Length (English)3.75 in.
Width (Metric)7.62cm
Width (English)3 in.
Dimensions (L x W x H)3.75 x 3 x 3.25 in. (9.52 x 7.62 x 8.25cm)
MaterialCopolyester Plastic
Includes4 insert to allow opening of 1, 2, 5 and 10mL ampules
For Use With (Application)Box style design for one handed opening of up to 3 ampules at a time, transparency allows unobstructed view at all times
Unit SizeEA