Sterile Graduated Transfer Pipets
Sterile Graduated Transfer Pipets
Actual product may vary
Sterile Graduated Transfer Pipets
Made up of low density polyethylene.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Length (Metric)Volume (Metric) DispensePackaging Type
FKS134891081 mL150 mm33 μLIndividually Wrapped
FKS134191181 mL150 mm33 μL50 inner packs of 10
FKS134691083 mL155 mm40 μLIndividually Wrapped
FKS134091183 mL155 mm40 μL25 inner packs of 20
FKS134791083 mL155 mm40 μL50 inner packs of 10
FKS134291181 mL150 mm33 μL25 inner packs of 20
Catalog number FKS13489108
Price (SGD)
75.00
PK
Add to cart
Volume (Metric):
1 mL
Length (Metric):
150 mm
Volume (Metric) Dispense:
33 μL
Packaging Type:
Individually Wrapped
Price (SGD)
75.00
PK
Add to cart
  • Transparent in color
  • Contains graduations
  • Sterile
Specifications
MaterialLDPE
Drops/mL30
GraduatedYes
Graduations1 mL
Quantity500/Pk.
Volume (Metric) Dispense33 μL
Packaging TypeIndividually Wrapped
Volume (Metric)1 mL
Length (Metric)150 mm
TypeTransfer Pipet
Unit SizePK