Write-On™ Paper Label Tapes
Write-On™ Paper Label Tapes
Actual product may vary
Write-On™ Paper Label Tapes
Write-On™ Paper Label Tapes
Fisherbrand
Write-On™ Paper Label Tapes
Use pencil, ball-point pen, or solvent ink marker to create labels that remain legible under the most demanding laboratory conditions. Paper label tapes resist moisture and most solvents and are great for use in low temperature applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorDimensions (LxW)Diameter (English)
FKS13455246Yellow36.4 m x 25.4 mm3 in.
FIS03-411-621Blue36.4 m x 12.7 mm3 in.
FIS03-411-607Green13.7 m x 19 mm1 in.
FIS03-411-622Blue36.4 m x 19 mm3 in.
FIS03-411-616Yellow36.4 m x 19 mm3 in.
FIS03-411-601White13.7 m x 19 mm1 in.
FIS03-411-619Green36.4 m x 19 mm3 in.
FIS03-411-600White13.7 m x 12.7 mm1 in.
FIS03-411-628White54.6 m x 19 mm3 in.
FIS03-411-625Orange36.4 m x 19 mm3 in.
FIS03-411-610White36.4 m x 19 mm3 in.
FIS03-411-604Yellow13.7 m x 19 mm1 in.
FIS11-865-109White, Yellow, Green13.7 m x 1.9 cm1 in.
FIS03-411-613Red36.4 m x 19 mm3 in.
FKS13434720Yellow36.4 m x 12.7 mm3 in.
Catalog number FKS13455246
Price (SGD)
306.00
PK
Add to cart
Color:
Yellow
Dimensions (LxW):
36.4 m x 25.4 mm
Diameter (English):
3 in.
Price (SGD)
306.00
PK
Add to cart
Use pencil, ball-point pen, or solvent ink marker to create labels that remain legible under the most demanding laboratory conditions. Paper label tapes resist moisture and most solvents and are great for use in low temperature applications.
  • Adheres to any clean surface, including PTFE fluoropolymer resin coated materials
  • Peels off without leaving a sticky residue
  • Pencil, ball-point pen, or solvent ink marker remain legible under the most demanding laboratory conditions
  • Colored paper with rubber resin adhesive resists moisture and most solvents
  • Withstands a maximum temperature of 157°C (315°F) for 30 minutes or 80°C (176°F) for 24 hours; can be used down to -80°C (-112°F)
  • Withstands a steam autoclaving cycle at 121°C (250°F) and gamma irradiation between 13.0 and 45.0kGy
Specifications
AutoclavableAutoclavable
Adhesive TypeRubber, Resin
ColorYellow
FinishMatte
FormatRoll
MaterialPaper
Printable SidesSingle-side Printable
Print TechnologyInk Jet
Product TypeUltra Reflective Letter Label
SterilityNon-sterile
For Use With (Application)For use in low temperature applications
Quantity3/Pk.
Dimensions (LxW)36.4 m x 25.4 mm
Length (English)40 yd.
Length (Metric)36.5 m
Width (English)1 in.
Width (Metric)25.4 mm
Diameter (English)3 in.
Diameter (Metric)7.62 cm
Unit SizePK