Trigger Sprayer, 53 mm Adapter
Trigger Sprayer, 53 mm Adapter
Actual product may vary
Fisherbrand
Trigger Sprayer, 53 mm Adapter
Trigger sprayers with adapters fit any Bel-Art bottle with an easy-to-fill, 53 mm-wide closure. They offer convenient, adjustable spray dispensing of a wide range of chemicals, including bleach, ethanol, machine oil and water.
Have Questions?
Catalog NumberdimCapacityMetricDiameter (Metric)Closure Size
FKS1343251953 mm53 mm
Catalog number FKS13432519
Price (SGD)
185.00
PK
Add to cart
Diameter (Metric):
53 mm
Closure Size:
53 mm
Price (SGD)
185.00
PK
Add to cart
Trigger sprayers with adapters fit any Bel-Art bottle with an easy-to-fill, 53 mm-wide closure. They offer convenient, adjustable spray dispensing of a wide range of chemicals, including bleach, ethanol, machine oil and water.
  • Simple-to-install sprayer is perfect for OSHA-compliant 16 oz. and 32 oz. Bel-Art™ non-vented, wide-mouth wash bottles (series 11716 and 11732).
  • Stainless steel, spring-action pump in the trigger sprayer will not corrode with repeated exposure to 10% bleach disinfecting solution.
  • Made from durable, chemical-resistant polypropylene.
  • Sprayer without adapter fits most bottles with a 28 mm opening.
  • Sprayer with adapter adds approx. 5.5 in. height to a bottle.
  • Comes in pack of 2.
  • Bottle not included.
Specifications
ColorWhite, Gray
Includes53mm Adapter, Stainless Steel spring action pump. Bottle not included.
MaterialPolypropylene
Quantity2/Pk.
SterilityNon-sterile
Closure IncludedYes
AutoclavableNot Autoclavable
DisposableReusable
GraduatedNo
MouthWide, Threaded
ShapeCylindrical
Height (Metric)139.7 mm
Product TypeTrigger Spray Head
Width (English)3 in.
Length (English)12 in.
Height (English)5.5 in.
Chemical LabelBlank
Closure MaterialPolypropylene
Closure Size53 mm
Diameter (Metric)53 mm
Closure TypeThreaded
Unit SizePK