Square and Rectangular Cover Glasses
Square and Rectangular Cover Glasses
Actual product may vary
Fisherbrand
Square and Rectangular Cover Glasses
Provides quality, reliability and performance for your laboratory microscopy needs
Have Questions?
Change viewbuttonViewtableView
Catalog NumberThickness (Metric)Length (Metric)Width (Metric)Shape
FKS13206778#1 (0.13 to 0.16 mm)22 mm22 mmSquare
FIS12-553-454#1.5 (0.16 to 0.19 mm)22 mm22 mmSquare
FIS12-553-465#1 (0.13 to 0.16 mm)60 mm24 mmRectangle
FIS12-553-450#1 (0.13 to 0.16 mm)18 mm18 mmSquare
Catalog number FKS13206778
Price (SGD)
610.00
PK
Add to cart
Thickness (Metric):
#1 (0.13 to 0.16 mm)
Length (Metric):
22 mm
Width (Metric):
22 mm
Shape:
Square
Price (SGD)
610.00
PK
Add to cart
Provides quality, reliability and performance for your laboratory microscopy needs
  • Made from soda-lime glass and borosilicate glass with high clarity
  • Available in square and rectangular shapes, and in three standard sizes and thicknesses
Specifications
Product TypeCoverslip
ShapeSquare
MaterialBorosilicate Glass
Length (Metric)22 mm
Width (Metric)22 mm
Thickness (Metric)#1 (0.13 to 0.16 mm)
SterilityNon-sterile
For Use With (Equipment)Microscopes
Quantity200/Pk.
Unit SizePK