Rotor for Centrifuge 5430 / 5430 R
Rotor for Centrifuge 5430 / 5430 R
Actual product may vary
Fisherbrand
Rotor for Centrifuge 5430 / 5430 R
Eppendorf Rotor for Centrifuge 5430/5430 R, precision at its core. Maximize your centrifuge's potential with rotors crafted for efficiency. Tailor your experiments with various capacities and speeds, ensuring optimal sample processing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityMax. RCFMax. SpeedIncludes
FKS1320658916 x 5.0 mL Eppendorf Tubes21,191 x g14,200 rpmAerosoal Tight Rotor Lid
FKS103685226 x 15/50 mL Conical or 12 x Vacutainer Tubes7,745 x g7,830 rpmRotor Lid
Catalog number FKS13206589
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
16 x 5.0 mL Eppendorf Tubes
Max. RCF:
21,191 x g
Max. Speed:
14,200 rpm
Includes:
Aerosoal Tight Rotor Lid
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight and PTFE coated options available.
Specifications
DescriptionFA-45-16-17, Aerosol-tight - includes rotor lid
MaterialAluminum
TypeFixed Angle Rotor
Capacity16 x 5.0 mL Eppendorf Tubes
Max. RCF21,191 x g
Max. Speed14,200 rpm
Max. Tube Diameter17 mm
For Use With (Equipment)Centrifuge 5430 / 5430 R
IncludesAerosoal Tight Rotor Lid
No. of Places16
Unit SizeEA