Duran Borosilicate Glass Lab Bottle
Actual product may vary
Fisherbrand
Duran Borosilicate Glass Lab Bottle
Duran Borosilicate glass lab bottle, pressure plus+. Clear, with DIN 168-1 thread.
Have Questions?
Catalog NumberCapacity (Metric)
FKS13197246500 mL
Catalog number FKS13197246
Price (SGD)
-
Capacity (Metric):
500 mL
Duran Borosilicate glass lab bottle, pressure plus+, Clear, with DIN 168-1 thread, GL 45.
Pressure and vacuum resistant glass laboratory bottles with plastic safety coating.
  • The DURAN™ pressure plus+ GL 45 protect coated bottles offer a safer option for laboratory applications involving a vacuum or pressure.
  • The coating provides scratch, leak*, and splinter* protection (* only applies to bottles 5000 ml and less).
  • Guaranteed vacuum and pressure resistant from –1 bar to + 1.5 bar due to modified geometry (based on ISO 4796-1).
  • Optimum safety for working under pressure or vacuum.
  • Suitable for full vacuum ps = -100 kPa.
  • Pressure resistance conforms to DIN EN 1595.
  • Fired-on, highly durable blue ceramic for visual differentiation from the standard laboratory bottle.
  • Easy-to-read scale with graduation marks to estimate the content.
Specifications
Capacity (Metric)500 mL
ColorClear
IncludesWithout discharge ring and dust protection cover
MaterialBorosilicate Glass
Quantity10 Pk.
Thread Size (Metric)GL 45
Certifications/ComplianceISO 4796-1
DisposableReusable
GraduatedYes
Diameter (Metric)86 mm