U-Tube Reverse Flow Viscometers
Actual product may vary
Fisherbrand
U-Tube Reverse Flow Viscometers
Suitable for transparent and opaque liquids. With UKAS certificate of calibration
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FKS131810331
FKS131710334
Catalog number FKS13181033
Price (SGD)
1,178.00
Each
Size:
1
Price (SGD)
1,178.00
Each
Type BS/IP/RF, IP71, IP319, ASTM D2170.Suitable for transparent and opaque liquids. Nominal overall length 275mm. Approximate sample size 12mL to 25mL. With UKAS certificate of calibration.
Specifications
Product TypeU-Tube Reverse Flow Viscometers
Viscosity Range0.6 to 3 cSt
ASTM NumbersD446
Certifications/ComplianceUKAS
Size1
Volume (Metric)12 to 25 mL
Unit SizeEach