DURAN™ Premium Screw Cap, Colourless PFA, with PTFE-coated (Platinum cured) silicone cap liner
DURAN™ Premium Screw Cap, Colourless PFA, with PTFE-coated (Platinum cured) silicone cap liner
Actual product may vary
DURAN™ Premium Screw Cap, Colourless PFA, with PTFE-coated (Platinum cured) silicone cap liner
DURAN™ Premium Screw Cap, Colourless PFA, with PTFE-coated (Platinum cured) silicone cap liner
Fisher Catalogue
DURAN™ Premium Screw Cap, Colourless PFA, with PTFE-coated (Platinum cured) silicone cap liner
Screw bottle closure with cap liner sealing that offers higher temperature and chemical resistance.
Have Questions?
Catalog NumberThread Size (Metric)
FKS13177226GL 25
Catalog number FKS13177226
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Thread Size (Metric):
GL 25
Thanks to its highly resistant materials, the premium cap is optimally suited to utilize in processes which are both chemically and thermally demanding. Due to its high temperature resistance, it is ideal for demanding processes, such as hot-air / dry-heat sterilisation and depyrogenation at 200°C. The materials used meet the requirements of the FDA/USP/EP and JP. Consequently the premium cap is particularly suitable for use in the pharmaceutical industry. Due to the PTFE-coated silicone cap liner, the premium cap provides a reliable and tight seal. Therefore it is ideal for the long term storage and transport of sensitive products. The matching PFA pouring ring permits drip-free pouring is available separately. Replacement PTFE faced silicone cap liners are available (platinum catalysed silicone)
  • High purity polymer without any colourants
  • USP Class VI Pharmaceutical grade materials
  • Fluoropolymer cap offers high temperature resistance
  • Fluoropolymer cap offers excellent chemical resistance
  • High tightness due to PTFE-coated silicone cap liner seal
  • Fully compatible with GL 25 or 45 threads and therefore highly versatile
  • The matching PFA pouring ring permits drip-free pouring and replacement PTFE faced silicone cap liners (platinum-cured silicone) are available separately
Specifications
DescriptionScrew cap
For Use With (Application)Autoclaving of media and reagents; Preparation of media and reagents; Reservoir cap for supplying laboratory instruments; Port assemblies for spinner flasks, and cell culture vessels; Single port vent for autoclaving media in large bottles; Transport and shipping
MaterialPFA, PTFE, Silicone
Quantity5/Pk.
Thread Size (Metric)GL 25
AutoclavableAutoclavable
Diameter (Metric)32 mm
Unit SizePK