Heated Timed Ultrasonic Bath with Drain Outlet
Heated Timed Ultrasonic Bath with Drain Outlet
Actual product may vary
Fisherbrand
Heated Timed Ultrasonic Bath with Drain Outlet
Get perfect and even distribution of ultrasonic activity. Designed for demanding applications and for continuous operation, industrial strength Clifton™ Heated Timed Ultrasonic Baths feature eco and dynamic sonic modes. Eco offers quieter ultrasonic activity, while dynamic provides vigorous cleaning. Complete with a basket and lid.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Model
FKS1316131330 LSW30H
FKS1315131312 LSW12H
Catalog number FKS13161313
Price (SGD)
13,128.00
PK
Add to cart
Capacity (Metric):
30 L
Model:
SW30H
Price (SGD)
13,128.00
PK
Add to cart
  • High performance industrial strength ultrasonics with sweep function for perfect and even distribution of ultrasonic activity
  • Stainless steel construction
  • Auto-degassing function useful for liquid changes in the tank, decarbonating and degassing HPLC solutions
  • Ultrasonic boost function for 25% more sonic power
  • Easy drain facility on 6L and above baths
  • Heaters can safely run dry
  • Temperature controlled ultrasonics - activity will start once set temperature is reached
  • Indication of time remaining
Specifications
MaterialStainless Steel
Timer1 to 30 min.
DisplayAnalog
Frequency38 kHz
Voltage120/230 V
Capacity (Metric)30 L
Depth (Metric)340 mm
Depth (Metric) Interior300 mm
Dimensions (D x W x H)340 x 568 x 321 mm
Dimensions (D x W x H) Interior200 x 505 x 300 mm
Electrical Requirements230/120 V, 38 kHz, 300 W
For Use With (Application)Designed for Demanding Applications and Suitable for Continuous Operation
Height (Metric) Interior200 mm
ModelSW30H
Temperature (Metric) Range30°C to 80°C
Wattage300 W
Width (Metric)568 mm
Width (Metric) Interior505 mm
Warranty3 Years
Unit SizePK