Sterile Sampling Bags with Flat-Wire Closures
Sterile Sampling Bags with Flat-Wire Closures
Actual product may vary
Fisherbrand
Sterile Sampling Bags with Flat-Wire Closures
An economical and efficient way to collect, contain and carry samples
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Thickness (English)ColorCapacity (English)
FKS131581679 x 5.5 in. (22.9 x 14cm)3.50 milClear24 oz.
FIS14-955-1869 x 4.5 in. (22.9 x 11.4cm)3 milClear18 oz.
FIS14-955-1879 x 5.5 in. (22.9 x 14cm)3.50 milClear24 oz.
FIS14-955-18815 x 5.5 in. (38.1 x 14cm)3.50 milClear42 oz.
FIS14-955-1899 x 4.5 in. (22.9 x 11.4cm)3 milClear with White Block18 oz.
FIS14-955-19510 x 10 in.4 milClear with White Block60 oz.
FIS14-955-19614 x 10 in. (35.6 x 25.4cm)4 milClear100 oz.
FIS14-955-19312 x 7 in. (30 x 18cm)3.50 milClear55 oz.
Catalog number FKS13158167
Price (SGD)
226.00
PK
Add to cart
Dimensions (L x W):
9 x 5.5 in. (22.9 x 14cm)
Thickness (English):
3.50 mil
Color:
Clear
Capacity (English):
24 oz.
Price (SGD)
226.00
PK
Add to cart
Collect, contain and carry samples in an economical and efficient way
  • Sterile to avoid sample contamination
Specifications
Capacity (English)24 oz.
Closure TypeFlat Wire
ColorClear
MaterialPolyethylene
Thickness (English)3.50 mil
Dimensions (L x W)9 x 5.5 in. (22.9 x 14cm)
Length (English)9 in.
Length (Metric)22.9 cm
Width (English)5.5 in.
Width (Metric)14 cm
Unit SizePK