WS-100 Series Stuart™ Water Still
WS-100 Series Stuart™ Water Still
Actual product may vary
Fisherbrand
WS-100 Series Stuart™ Water Still
Save money while maintaining high performance with this WS-100 Series Stuart Water Still.
Have Questions?
Catalog NumberVoltage
FKS13145990240 V
Catalog number FKS13145990
Price (SGD)
5,093.00
EA
Add to cart
Voltage:
240 V
Price (SGD)
5,093.00
EA
Add to cart
  • Two independent thermostats prevent overheating.
  • Features a sturdy metal stand for either wall or bench mounting.
  • Operate directly from tap water.
  • This basic economical water still is perfect for general laboratory and educational distillations.
  • Borosilicate glass boiler and condenser provide the highest purity distillate.
  • The unique condenser design ensures that the distilled water droplets remain in contact with the cooling coil for an extended period for maximum energy transfer and immediate cold distilled water.
  • Condenser with screw thread connections and plastic 3/8 in. ID hose barb fittings allow quick and safe tubing change without worrying about breaking the glassware.
  • Easily descale without dismantling the glassware using the built-in acid addition funnel and wastewater drain.
  • Long-life chromium-plated metal heater is protected by twin safety thermostats.
Specifications
Electrical Requirements220/240 V 50/60 Hz 3 kW
Frequency50/60 Hz
Phase1
Voltage240 V
Inlet Water Temperature (Metric)25°C to 35°C
TypeMerit Water Still
Output Flow Rate4 L/hr.
ModelW4000
Depth (English)5.9 in.
Depth (Metric)150 mm
Height (English)17.7 in.
Height (Metric)450 mm
Width (English)19.7 in.
Width (Metric)500 mm
Unit SizeEA