Kink-resistant for maximum flexibility, with a smooth bore for ease of cleaning
Have Questions?
Catalog Number
Diameter (Metric) Inner
FKS13140873
4.75 mm
Catalog number FKS13140873
Price (SGD)
-
Diameter (Metric) Inner:
4.75 mm
Flexible, highly elastic but kink-resistant for maximum versatility. Grips tightly to metal or glass to provide secure connections. Smooth bore, for ease of cleaning. Non-toxic.
Specifications
Diameter (Metric) Inner4.75 mm
MaterialPVC
Diameter (Metric) Outer7.92 mm
Overall Length (Metric)30 m
For Use With (Application)To provide secure connections