Micromeg Disposable Replacement Cartridge
Micromeg Disposable Replacement Cartridge
Actual product may vary
Fisherbrand
Micromeg Disposable Replacement Cartridge
For the production of low volumes of deionised water.
Have Questions?
Catalog number FKS13135940
Price (SGD)
4,607.00
PK
Add to cart
Price (SGD)
4,607.00
PK
Add to cart
  • Produces water up to a quality of 10MΩ/cm at flows of up to 90L/hr
  • Requires no electrical input
  • Cartridges can be replaced quickly
Specifications
Flow Rate90 L/hr.
Filter SizeMC:DW cartridges
For Use With (Equipment)Micromeg Disposable Deionizer PRDIDC0020
Inlet Pressure3.4 bar
Unit SizePK