Freeze Drying Ampules
Freeze Drying Ampules
Actual product may vary
Fisherbrand
Freeze Drying Ampules
Use these ampules for a variety of laboratory applications. Freeze Drying Ampules are manufactured neutral glass to the dimensions, tolerances and quality requirements of ISO 4142:2002, to meet all applications for re-usable glass tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS131317932.5 mL
FKS131217930.5 mL
Catalog number FKS13131793
Price (SGD)
797.00
PK
Add to cart
Capacity (Metric):
2.5 mL
Price (SGD)
797.00
PK
Add to cart
Use these ampules for a variety of laboratory applications. Freeze Drying Ampules are manufactured neutral glass to the dimensions, tolerances and quality requirements of ISO 4142:2002, to meet all applications for re-usable glass tubes.
Clear neutral glass vials for freeze drying. The top 11mm of the open ends have internal neck diameter 5.9mm to 6.1mm and are glazed. Capacity and length are nominal.
Specifications
Capacity (Metric)2.5 mL
MaterialNeutral Glass
Product TypeAmpule
ColorClear
Diameter (Metric) Outer7 mm
Length (Metric)154mm
Unit SizePK