Heated Timed Ultrasonic Bath
Heated Timed Ultrasonic Bath
Actual product may vary
Fisherbrand
Heated Timed Ultrasonic Bath
Get perfect and even distribution of ultrasonic activity. Designed for demanding applications and for continuous operation, industrial strength Clifton™ Heated Timed Ultrasonic Baths feature eco and dynamic sonic modes. Eco offers quieter ultrasonic activity, while dynamic provides vigorous cleaning.
Have Questions?
Catalog NumberCapacity (Metric)Model
FKS131313133 LSW3H
Catalog number FKS13131313
Price (SGD)
3,692.00
EA
Add to cart
Capacity (Metric):
3 L
Model:
SW3H
Price (SGD)
3,692.00
EA
Add to cart
  • High performance industrial strength ultrasonics with sweep function for perfect and even distribution of ultrasonic activity
  • High power piezoelectric transducers
  • Stainless steel construction
  • Auto-degassing function useful for liquid changes in the tank, decarbonating and degassing HPLC solutions
  • Ultrasonic boost function for 25% more sonic power
  • Heaters can safely run dry
  • Temperature controlled ultrasonics - activity will start once set temperature is reached
  • Indication of time remaining
  • Recommended accessories: Lid and Basket are not included; sold separately
Specifications
MaterialStainless Steel
Timer1 to 30 min.
DisplayAnalog
Frequency38 kHz
Voltage120/230 V
Capacity (Metric)3 L
Depth (Metric)150 mm
Depth (Metric) Interior137 mm
Dimensions (D x W x H)150 x 250 x 220 mm
Dimensions (D x W x H) Interior100 x 240 x 137 mm
Electrical Requirements230/120 V, 38 kHz, 200 W
For Use With (Application)Designed for Demanding Applications and Suitable for Continuous Operation
Height (Metric) Interior100 mm
ModelSW3H
Temperature (Metric) Range30°C to 80°C
Wattage200 W
Width (Metric)250 mm
Width (Metric) Interior240 mm
Warranty3 Years
Unit SizeEA