Check Order Status
Replacement Pen Cartridge
Actual product may vary
Fisherbrand
Replacement Pen Cartridge
Sound and count occur simultaneously when inserted correctly into Counter-Pen™. Fisherbrand™ Replacement Pen Cartridge marks with black felt tip.
Have Questions?
Catalog Number
Color
FKS13128325
Black
Catalog number
FKS13128325
Price (SGD)
34.00
EA
Add to cart
Color:
Black
Price (SGD)
34.00
EA
Add to cart
Sound and count occur simultaneously when inserted correctly into Counter-Pen™. Fisherbrand™ Replacement Pen Cartridge marks with black felt tip.
Specifications
Product Type
Replacement Cartridge for Counter-pen
Tip Style
Felt Tip
Color
Black
For Use With (Equipment)
Counter-Pen™