Cyclon Ultrapure Water Still
Cyclon Ultrapure Water Still
Actual product may vary
Fisherbrand
Cyclon Ultrapure Water Still
Highly suitable still for life science applications. Fistreem International™ Cyclon Ultrapure Water Still employs modern electronics and dual safety systems to produce high-quality, pyrogen-free distillate.
Have Questions?
Catalog NumberOutput Flow RateVoltageDimensions (L x W x H)
FKS131160104 L/hr. Single Distilled240 V300 x 380 x 750 mm
Catalog number FKS13116010
Price (SGD)
22,379.00
EA
Add to cart
Output Flow Rate:
4 L/hr. Single Distilled
Voltage:
240 V
Dimensions (L x W x H):
300 x 380 x 750 mm
Price (SGD)
22,379.00
EA
Add to cart
  • Designed to surpass accepted purity and performance levels of traditional systems.
  • Of particular interest to users in life sciences, the pyrogen free distillate is of the very highest purity.
  • Unique condenser vapor trap creates a cyclonic effect eliminating any vapor borne droplets, thereby preventing carry-over in the distillate.
  • Outputs of 4L/hr and 8L/hr single distilled and 4L/hr double distilled are offered.
  • High quality borosilicate glassware houses very durable, silica sheathed, heating elements.
  • Protection of the heaters by thermistor control and a thermal fuse allows unattended safe operation.
  • Auto reservoir controls automatically switch off the still on a full reservoir.
  • Supplied ready for connection to a mains tap supply and will provide high quality pyrogen free water, even on the hardest feed water supplies. Cyclon deionizer is specially designed for use with the still.
  • Boiler contents are drained automatically on switch off to reduce scale formation rate.
  • May be fed with a suitable quality pre-treated water to reduce or eliminate the need for periodic cleaning.
    • In this condition the condenser water (tap feed) is separated from the boiler water (treated feed) for economy.
    • Supplied with pre-treated feed components that are easily fitted to allow treated water to be delivered directly to the boiler.
    • In this condition controls ensure that the treated feed water is automatically supplied on demand to avoid any wastage.
    • Suitable feed water should be deionized, reverse osmosis or distilled quality.
  • A low water pressure adapter is available for use with feed water pressure between 0.35bar and 0.70bar (5psi to 10psi).
Specifications
Dimensions (L x W x H)300 x 380 x 750 mm
Electrical Requirements240 V 3 kW
Voltage240 V
TypeCyvlon Still
Output Flow Rate4 L/hr. Single Distilled
Depth (Metric)380 mm
Height (Metric)750 mm
Width (Metric)300 mm
Unit SizeEA