CD115 (c-fms) Monoclonal Antibody (12-3A3-1B10), Biotin, eBioscience™, Invitrogen™
CD115 (c-fms) Monoclonal Antibody (12-3A3-1B10), Biotin, eBioscience™, Invitrogen™
Actual product may vary
Fisherbrand
CD115 (c-fms) Monoclonal Antibody (12-3A3-1B10), Biotin, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Have Questions?
Catalog NumberQuantity
FKS1311598025 μg
Catalog number FKS13115980
Price (SGD)
1,934.00
EA
Add to cart
Quantity:
25 μg
Price (SGD)
1,934.00
EA
Add to cart
Description: CD115 or Colony-Stimulating Factor-1 Receptor (CSF-1R) is expressed on cells committed to the monocyte lineage and osteoclasts. CSF-1R is a disulfide-linked homodimer, which upon binding of its ligand, CSF-1, undergoes tyrosine autophosphorylation and subsequently, phosphorylates other membrane-proximal downstream targets which results in cytoskeletal remodeling, gene transcription and protein translation. CSF-1R activation promotes the survival, proliferation and differentiation of mononuclear phagocytes and the spreading and motility of macrophages. In osteoclasts, CSF-1R synergizes with RANKL to regulate the differentiation of mononuclear phagocytes to osteoclasts. Applications Reported: This 12-3A3-1B10 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 12-3A3-1B10 antibody has been tested by flow cytometric analysis of normal human peripheral blood. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
CSF1R (FMS) is involved in the production, differentiation, and function of macrophages, and is implicated in promyelocytic leukemias. Ligand binding activates the receptor kinase through a process of oligomerization and transphosphorylation. CSF1R is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases. Mutations in the gene encoding CSF1R have been associated with a predisposition to myeloid malignancy.Tyrosine-protein kinase that acts as cell-surface receptor for CSF1 and IL34 and plays an essential role in the regulation of survival, proliferation and differentiation of hematopoietic precursor cells, especially mononuclear phagocytes, such as macrophages and monocytes. Promotes the release of proinflammatory chemokines in response to IL34 and CSF1, and plays an important role in innate immunity and in inflammatory processes. The first intron of the CSF1R gene contains a transcriptionally inactive ribosomal protein L7 processed pseudogene oriented in the opposite direction. Mutations in the CSF1R gene have been associated with a predisposition to myeloid malignancy.
For Research Use Only.
Specifications
AntigenCD115 (c-fms)
ApplicationsFlow Cytometry
ClassificationMonoclonal
Clone12-3A3-1B10
Concentration0.5 mg/mL
ConjugateBiotin
FormulationPBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
GeneCSF1R
Gene Accession No.P07333
Gene AliasC-FMS, CD115, CSFR, FIM2, FMS
Gene SymbolsCSF1R
Host SpeciesRat
Purification MethodAffinity chromatography
Quantity25 μg
Regulatory StatusRUO
Primary or SecondaryPrimary
Gene ID (Entrez)1436
Target SpeciesHuman
Product TypeAntibody
FormLiquid
IsotypeIgG2a κ
Unit SizeEA
Contents & Storage
4° C, store in dark, DO NOT FREEZE!