Osmometer Calibration Standards
Actual product may vary
Osmometer Calibration Standards
Calibration standards for use with Advanced osmometers
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentrationFor Use With (Equipment)Quantity
FKS1310209350 mOsm/kg2020, 3300 and 3320 Osmometers10 x 2 mL
FKS12812201850 mOsm/kg2020, 3300 and 3320 Osmometers10 x 2 mL
Catalog number FKS13102093
Price (SGD)
-
Concentration:
50 mOsm/kg
For Use With (Equipment):
2020, 3300 and 3320 Osmometers
Quantity:
10 x 2 mL
Calibration standards for use with Advanced osmometers
  • Range from 50 to 3000 mOsm/kg
Specifications
ColorClear
Concentration50 mOsm/kg
For Use With (Equipment)2020, 3300 and 3320 Osmometers
TypeOsmometer Calibration Standard
IncludesSodium Chloride 0.15% to 9.5%, 2-Hydroxybiphenyl Sodium Salt Tetrahydrate <0.1%
Intended UseOsmometer instrument calibration and quality control
Quantity10 x 2 mL
FormLiquid
Unit SizePack
Contents & Storage
Room Temperature