Clifton™ Stainless Steel Basket
Clifton™ Stainless Steel Basket
Actual product may vary
Fisherbrand
Clifton™ Stainless Steel Basket
Complete an Ultrasonic Bath with a stainless steel basket. Nickel Electro™ Clifton™ Stainless Steel Basket is durable and made of high quality stainless steel.
Have Questions?
Catalog NumberCapacity (Metric)Dimensions (L x W x H)
FKS131013233 L198 x 106 x 50 mm
Catalog number FKS13101323
Price (SGD)
415.00
EA
Add to cart
Capacity (Metric):
3 L
Dimensions (L x W x H):
198 x 106 x 50 mm
Price (SGD)
415.00
EA
Add to cart
  • High quality
  • Durable
  • Stainless steel
  • For use with Ultrasonic Bath
  • Includes side lifting handles
Specifications
TypeBasket
Capacity (Metric)3 L
Depth (Metric)106 mm
For Use With (Application)For Ultrasonic Bath
Height (Metric)50 mm
MaterialStainless Steel
Width (Metric)198 mm
Dimensions (L x W x H)198 x 106 x 50 mm
Unit SizeEA