1x1 image pixel for data collection
Total Immersion Pocket Thermometer
Total Immersion Pocket Thermometer
Actual product may vary
Fisherbrand
Total Immersion Pocket Thermometer
Brannan Total Immersion Pocket Thermometer, Temperature Range: -10 deg. to 110 deg.C, Bottle Fill: Red spirit, Immersion Type: Total Immersion, Includes: Protective case and pocket clip, Length: 155mm, Max. Temperature: 110 deg.C, Min. Temperature: -10 deg.C, Width: 15mm
Have Questions?
Catalog NumberTemperature Range (Metric)
FKS13099212-10°C to +110°C
Catalog number FKS13099212
Price (SGD)
-
Temperature Range (Metric):
-10°C to +110°C
Brannan Total Immersion Pocket Thermometer, Temperature Range: -10 deg. to 110 deg.C, Bottle Fill: Red spirit, Immersion Type: Total Immersion, Includes: Protective case and pocket clip, Length: 155mm, Max. Temperature: 110 deg.C, Min. Temperature: -10 deg.C, Width: 15mm
Specifications
Bottle FillRed Spirit
Immersion TypeTotal Immersion
IncludesProtective case and pocket clip
Length (Metric)155 mm
Product TypePocket Thermometer
Max. Temperature (English)230°F
Max. Temperature (Metric)110°C
Min. Temperature (English)14°F
Min. Temperature (Metric)-10°C
Temperature Range (Metric)-10°C to +110°C
Width (Metric)15 mm
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link