Enviro-Safe™ General Purpose Liquid-In-Glass Thermometers, Partial Immersion 3 in.
Actual product may vary
Fisherbrand
Enviro-Safe™ General Purpose Liquid-In-Glass Thermometers, Partial Immersion 3 in.
Certified environmentally safe thermometers replace dangerous mercury
Have Questions?
Catalog NumberTemperature Range (Metric)TemperatureRangeEnglishScale
FKS13099132-10°C to +260°CCelsius
Catalog number FKS13099132
Price (SGD)
-
Temperature Range (Metric):
-10°C to +260°C
Scale:
Celsius
  • Earth friendly, EnviroKleen™ certified thermometers
  • Feature green, non-toxic, biodegradable, Enviro-Safe™ liquid against lead-free, white back glass
  • Meet government, education, and industry mandates to purchase “Green” products
Applications:
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Specifications
Certifications/ComplianceIndividually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
For Use With (Application)Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Immersion TypePartial
Length (English) Overall14 in.
Length (Metric) Overall350 mm
Max. Temperature (Metric)260°C
Min. Temperature (Metric)-10°C
Temperature Range (Metric)-10°C to +260°C
Accuracy±1.5°C, ±2°C above 100°C, ±2.5°C above 200°C, ±3°C above 230°C
Subdivision1°C
Immersion Length76 mm
ScaleCelsius
Unit SizePack