Hard Wood Test Tube Racks
Hard Wood Test Tube Racks
Actual product may vary
Fisherbrand
Hard Wood Test Tube Racks
Organize and store test tubes and samples with these stands. Hard Wood Test Tube Racks occupy only about one third of the space required by a conventional rack when dismantled for storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) Cavity
FKS1309842322 mm
FKS1302843332 mm
FKS1300843310 mm
Catalog number FKS13098423
Price (SGD)
168.00
PK
Add to cart
Diameter (Metric) Cavity:
22 mm
Price (SGD)
168.00
PK
Add to cart
  • Single row test tube stand
  • Hardwood top and bottom members
  • Molded polypropylene supports which clip into position
  • Packs flat for transport and storage
Specifications
Product TypeTest Tube Rack
Array6
HoldsTubes (22 mm)
MaterialHardwood
Positions6
Quantity5 Pack
Diameter (Metric) Cavity22 mm
Diameter (Metric) Opening22 mm
Unit SizePK