FRIO-Temp™ Glass Thermometers: Enviro-Safe™ Filled
FRIO-Temp™ Glass Thermometers: Enviro-Safe™ Filled
Actual product may vary
FRIO-Temp™ Glass Thermometers: Enviro-Safe™ Filled
FRIO-Temp™ Glass Thermometers: Enviro-Safe™ Filled
Fisher Catalogue
FRIO-Temp™ Glass Thermometers: Enviro-Safe™ Filled
Ideal for use in freezers, refrigerators, incubators, ovens and blood banks.
Have Questions?
Catalog NumberTemperature Range (Metric)For Use With (Application)Product Type
FKS1308916215°C to 50°CIncubatorEasy Read Verification Thermometer
Catalog number FKS13089162
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Temperature Range (Metric):
15°C to 50°C
For Use With (Application):
Incubator
Product Type:
Easy Read Verification Thermometer
Come complete with a Statement of Accuracy traceable to NIST™ and DKD, unique serial number, and Velcro™ and magnet attachments. Simply screw thermometer with cap into bottle and attach to test sample.
Specifications
Bottle Size30 mL
Certifications/ComplianceISO/IEC 17025:2005, A2LA, NIST, DAkkS
For Use With (Application)Incubator
IncludesPTFE coating
Length (English) Overall5.3 in.
Length (Metric) Overall135 mm
MaterialGlass
Thermometer FillEnviro-Safe™ Liquid
Product TypeEasy Read Verification Thermometer
Max. Temperature (Metric)50°C
Min. Temperature (Metric)15°C
Temperature Range (Metric)15°C to 50°C
Accuracy±0.5°C
Subdivision0.5°C
Unit SizeEA