General Purpose High Accuracy Lab Thermometers
Actual product may vary
Fisherbrand
General Purpose High Accuracy Lab Thermometers
A general purpose laboratory thermometer of high accuracy, mercury-filled and manufactured from yellow back glass.
Have Questions?
Catalog NumberTemperature Range (Metric)
FKS13088623-100°C to +50°C
Catalog number FKS13088623
Price (SGD)
-
Temperature Range (Metric):
-100°C to +50°C
  • Supplied in individual, resealable and telescopic tubes
  • Can be supplied with a UKAS calibration certificate or statement of conformity upon request
  • Total immersion thermometer
  • Temperature range of −100° to +50°C
Specifications
Bottle FillRed Spirit
Certifications/ComplianceUKAS calibration certificate, statement of conformity
For Use With (Application)General use
Immersion TypeTotal Immersion
IncludesIndividual, resealable storage tube
Length (Metric)305 mm
MaterialGlass
Product TypeLaboratory Thermometer
Max. Temperature (Metric)50°C
Min. Temperature (Metric)-100°C
Temperature Range (Metric)-100°C to +50°C
Subdivision1°C
Unit SizeEach