Lab Tripod Thermometer
Actual product may vary
Fisherbrand
Lab Tripod Thermometer
General purpose laboratory thermometer without calibration certificate.
Have Questions?
Catalog NumberTemperature Range (Metric)
FKS13078573-10°C to +110°C
Catalog number FKS13078573
Price (SGD)
92.00
Pack
Temperature Range (Metric):
-10°C to +110°C
Price (SGD)
92.00
Pack
Filled with a brightly colored, mercury-free spirit. Brannan™ Partial Immersion Spirit Filled Thermometers use specially selected filling materials covering temperatures down to -200°C and feature a non-roll cap with suspension ring.
  • Lab thermometer
  • Safe non-mercury filling
  • Non-roll end cap
  • Supplied as standard in protective individual, resealable telescopic tube
  • Made in UK
Specifications
For Use With (Application)Ideal for educational use
Immersion TypePartial Immersion
Length (Metric)305 mm
MaterialGlass
Thermometer FillGreen Kerosene Spirit
Weight (Metric)16.2 g
Product TypeLaboratory Thermometer
Max. Temperature (English)230°F
Max. Temperature (Metric)110°C
Min. Temperature (English)14°F
Min. Temperature (Metric)-10°C
Temperature Range (Metric)-10°C to +110°C
For Use With (Equipment)Educational Use
Accuracy±1.0 at 35°C
Subdivision1°C
Immersion Length76 mm
Width (Metric)1.2 cm
Unit SizePack