Check Order Status
Brass Test Tube Holder
Actual product may vary
Fisherbrand
Brass Test Tube Holder
Accepts tubes of up to 45mm in diameter.
Have Questions?
Catalog Number
Material
FKS13068423
Brass
Catalog number
FKS13068423
Price (SGD)
123.00
PK
Add to cart
Material:
Brass
Price (SGD)
123.00
PK
Add to cart
With a wooden handle
Overall length of 225mm
Excellent for general laboratory use
Specifications
For Use With (Application)
For 45mm dia. Tubes
Includes
with Wooden Handle
Quantity
1 Ea.
Length (Metric)
225 mm
Material
Brass
Type
Test Tube Holder