Pyrex™ Borosilicate Glass Medium Wall Rimless Test Tubes
Pyrex™ Borosilicate Glass Medium Wall Rimless Test Tubes
Actual product may vary
Pyrex™ Borosilicate Glass Medium Wall Rimless Test Tubes
Pyrex™ Borosilicate Glass Medium Wall Rimless Test Tubes
Fisherbrand
Pyrex™ Borosilicate Glass Medium Wall Rimless Test Tubes
Manufactured from Pyrex™ borosilicate glass to give superior performance over neutral borosilicate and soda lime glass. Pyrex™ Borosilicate Glass Medium Wall Rimless Test Tubes are ideal for microbiology culture work.
Have Questions?
Catalog NumberDiameter (Metric)Length (Metric)Capacity (Metric)
FKS1305839324 mm150 mm55 mL
Catalog number FKS13058393
Price (SGD)
676.00
PK
Add to cart
Diameter (Metric):
24 mm
Length (Metric):
150 mm
Capacity (Metric):
55 mL
Price (SGD)
676.00
PK
Add to cart
Manufactured from Pyrex™ borosilicate glass to give superior performance over neutral borosilicate and soda lime glass. Pyrex™ Borosilicate Glass Medium Wall Rimless Test Tubes are ideal for microbiology culture work.
  • Rimless medium walls: constructed for optimum performance
  • Robust construction: allows use at high temperatures up to 500deg.C
  • Conforms to: ISO 4142
Specifications
Certifications/ComplianceISO 4142
Capacity (Metric)55 mL
Length (Metric)150 mm
MaterialBorosilicate Glass
For Use WithIdeal for microbiology culture work
Quantity100 Pack
RimmedNo
Diameter (Metric)24 mm
Unit SizePK