Double-Safe™ PFA Safety Coated, Liquid-In-Glass Thermometers: Total Immersion
Double-Safe™ PFA Safety Coated, Liquid-In-Glass Thermometers: Total Immersion
Actual product may vary
Fisherbrand
Double-Safe™ PFA Safety Coated, Liquid-In-Glass Thermometers: Total Immersion
Protect your workplace and ensure a safe environment
Have Questions?
Catalog NumberTemperature Range (Metric)Temperature Range (English)Length (Metric) ProbeSubdivision
FKS13049152-20°C to +110°C300 mm1°C
Catalog number FKS13049152
Price (SGD)
-
Temperature Range (Metric):
-20°C to +110°C
Temperature Range (English):
Length (Metric) Probe:
300 mm
Subdivision:
1°C
  • Environmentally safe thermometers are PFA safety coated to keep liquid and glass contained in the event of breakage
  • Lead-free, yellow back glass and black liquid provides easy readability and fewer reading errors
  • Contains non-toxic, biodegradable, Enviro-Safe™ liquid
  • Manufactured and tested by our USA quality team
Applications:
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Specifications
Certifications/ComplianceTraceable to NIST. Includes a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
For Use With (Application)Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
GradePrecision
Immersion TypeTotal
Length (English) Overall12 in.
Length (Metric) Overall300 mm
Length (Metric) Probe300 mm
MaterialGlass
Thermometer FillEnviro-Safe™ Liquid
Max. Temperature (Metric)110°C
Min. Temperature (Metric)-20°C
Temperature Range (English)
Temperature Range (Metric)-20°C to +110°C
Accuracy±1°C
Subdivision1°C
Unit SizePK