Search Thermo Fisher Scientific
Catalog Number | Volume (Metric) | Quantity |
---|---|---|
FKS12999477 | 500/Pk., 3000/Cs. | |
FKS11349203 | 1 mL | 1/Pk., 500/Cs. |
FKS12909487 | 3 mL | 500/Pk., 3000/Cs. |
FKS11369203 | 3 mL | 10/Pk., 500/Cs. |
FKS11379203 | 3 mL | 1/Pk., 500/Cs. |
Ordering Information