MINO Series Classic Ovens
MINO Series Classic Ovens
Actual product may vary
Fisherbrand
MINO Series Classic Ovens
The simple to use thermostat system is good for teaching purposes and the units can be built up to meet individual requirements. Genlab™ MINO Series Classic Ovens are highly efficient, reliable and cost effective units to suit most drying, warming, sterilizing and general laboratory applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)No. of ShelvesWattage
FKS1299871818 L2500 w
FKS1293872875 L21000 w
Catalog number FKS12998718
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
18 L
No. of Shelves:
2
Wattage:
500 w
Design and Construction
  • Exterior is constructed from sheet steel finished in an easy clean powder coated paint
  • Interior chamber is made from mild steel coated with aluminum (CLAD)
  • Fitted with fixed shelf runners and removable chrome plated wire grid shelves
  • Solid steel door with controls located below the door
Heating and Controls
  • Heated by Incoloy sheathed elements positioned below the chamber floor
  • Easy to use thermostatic mechanical controls (range: 40 – 250°C)
  • Direct reading thermostat and overheat thermostat, both with calibrated scales and tamper proof locks
  • Includes main switch with indicator and heat and overheat indicators
Specifications
Certifications/ComplianceCE
Chamber MaterialSteel
Controller TypeMechanical thermostatic controls
Convection TypeNatural , using Incoloy sheathed elements
Capacity (Metric)18 L
Temperature (Metric)40°C to 250°C
TypeGeneral Purpose Convection Oven
DisplayDirect ReadingThermostat
For Use With (Application)Drying, warming, sterilizing and general laboratory applications
MaterialSteel
ModelMINO
No. of Shelves2
Safety FeaturesOverheat thermostat, tamper proof lock
Shelf MaterialRemovable chrome plated wire grid shelves
Temperature Stability±0.75°C
Warranty2 Years
Dimensions (D x W x H) Interior27 x 26 x 26 cm
Dimensions (L x W x H)38 x 41 x 52 cm
Length (Metric)38 cm
Width (Metric)41 cm
Height (Metric)52 cm
Weight (Metric)14 kg
Wattage500 w
Unit SizeEA