Vacutainer™ Glass Cell Preparation Tubes
Vacutainer™ Glass Cell Preparation Tubes
Actual product may vary
Fisherbrand
Vacutainer™ Glass Cell Preparation Tubes
Use for the one-step isolation of lymphocytes, monocytes, and plasma. BD™ Vacutainer™ Glass Cell Preparation Tubes allow for collection and processing of whole blood in a closed, sterile tube prepared with sodium citrate or sodium heparin as an anticoagulant.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure ColorAdditiveTube Size
FKS129976964 mLBlue/BlackSodium Citrate/FICOLL13 x 100 mm
FKS129177068 mLOrange/GreenSodium Heparin16 x 125 mm
FKS123792998 mLBlue/BlackSodium Citrate/FICOLL16 x 125 mm
Catalog number FKS12997696
Price (SGD)
3,590.00
PK
Add to cart
Capacity (Metric):
4 mL
Closure Color:
Blue/Black
Additive:
Sodium Citrate/FICOLL
Tube Size:
13 x 100 mm
Price (SGD)
3,590.00
PK
Add to cart
The BD CPT™ (Cell Preparation Tube) system provides a simple, standardized method for the isolation of Peripheral Blood Mononuclear Cells (PBMCs) from whole blood. In a single process, up to 15 million PBMCs can be isolated within 20 minutes.
  • Improved sample preparation consistency and reproducibility compared to manual gradient separations
  • Faster separations because there is no need to prepare gradients or manipulate the sample
  • Safe isolation of cells with reduced risk of contamination due to the enclosed, sterile tube
Vacutainer™ CPT™ features:
  • Glass molecular diagnostics tubes with see-thru label for easy content viewing
  • Conventional closures are color-coded to easily identify anticoagulant used
  • Density gradient polymer gel with either sodium citrate or sodium heparin anticoagulant
  • Blood draw capacity is 4 or 8mL (4mL tube available in sodium citrate only)
  • Tube is CE marked for in vitro diagnostic use
  • Centrifuging conditions: 1500–1800 g for 15 minutes at 18–25°C
Specifications
ColorClear
Product TypeCell Preparation Tube
AdditiveSodium Citrate/FICOLL
MaterialGlass
SterilitySterile
Closure TypeSpeckled Conventional
Capacity (Metric)4 mL
Closure ColorBlue/Black
Height (Metric)100 mm
Tube Size13 x 100 mm
IncludesPolyester gel, density gradient liquid
Label TypeTransparent
Diameter (Metric) Outer13 mm
Certifications/ComplianceCE
Unit SizePK