CG16K Capsule Dial Gauge
CG16K Capsule Dial Gauge
Actual product may vary
Fisherbrand
CG16K Capsule Dial Gauge
Barometrically compensated design gives better repeatability than bourdon gauges, especially at low pressures
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaximum Pressure
FKS129952740to 25 mbar
FKS129652740 to 1040 mbar
Catalog number FKS12995274
Price (SGD)
3,188.00
EA
Add to cart
Maximum Pressure:
0to 25 mbar
Price (SGD)
3,188.00
EA
Add to cart
Barometrically compensated design gives better repeatability than bourdon gauges, especially at low pressures. N. B. Use an adapter plus Co-Seal and clamp if retro fitting previous model CG3 with 3/8 inch BSP fitting. For adapter, please contact Customer Service.
  • Rugged gauge with easy to read 100mm dial
  • Accuracy ±2%
  • NW16 vacuum flange Mechanical gauge for use with pumps.
Specifications
TypeCapsule Dial Gauge
Maximum Pressure0to 25 mbar
Unit SizeEA